Assay Design
Gene Transcripts
- Gene Symbol
- bath-4
- Entrez Gene ID
-
186090
- Gene Name
- BTB and MATH domain containing
- Gene Aliases
-
CELE_F52C6.8
- Chromosome Location
-
Chr.II: 1916713 - 1917962 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061721.2 |
NP_494122.1 |
2 - 3 |
513 |
152 |
View Full Product Details