Assay Design
Gene Transcripts
- Gene Symbol
- fbxb-15
- Entrez Gene ID
-
173535
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_ZC204.7
- Chromosome Location
-
Chr.II: 1651836 - 1653497 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061625.5 |
NP_494026.1 |
2 - 2 |
773 |
137 |
View Full Product Details