Assay Design
Gene Transcripts
- Gene Symbol
- thn-1
- Entrez Gene ID
-
185060
- Gene Name
- THaumatiN family
- Gene Aliases
-
CELE_F28D1.3
- Chromosome Location
-
Chr.IV: 12380084 - 12380900 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069959.3 |
NP_502360.1 |
1 - 1 |
75 |
145 |
View Full Product Details