Assay Design
Gene Transcripts
- Gene Symbol
- grsp-4
- Entrez Gene ID
-
179146
- Gene Name
- Glycine Rich Secreted Protein
- Gene Aliases
-
CELE_F07C4.7
- Chromosome Location
-
Chr.V: 7624952 - 7626467 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072569.4 |
NP_504970.1 |
1 - 1 |
61 |
74 |
View Full Product Details