Assay Design



Gene Transcripts

Gene Symbol
srx-87
Entrez Gene ID
3565609
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F38B7.8
Chromosome Location
Chr.V: 11564873 - 11566771 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028726.1 NP_001023897.1 6 - 7 821 77

View Full Product Details