Assay Design
Gene Transcripts
- Gene Symbol
- tbc-10
- Entrez Gene ID
-
3564790
- Gene Name
- Rab-GAP TBC domain-containing protein
- Gene Aliases
-
CELE_R06B10.5
- Chromosome Location
-
Chr.III: 983261 - 988446 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064909.5 |
NP_497310.2 |
6 - 7 |
1250 |
82 |
View Full Product Details