Assay Design



Gene Transcripts

Gene Symbol
cpg-7
Entrez Gene ID
259477
Gene Name
Chondroitin proteoglycan 7
Gene Aliases
CELE_K09E4.6
Chromosome Location
Chr.II: 14108016 - 14109237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027072.4 NP_001022243.1 1 - 2 72 56
GenBank mRNA DQ340629.1 - 1 - 2 52 56

View Full Product Details