Assay Design



Gene Transcripts

Gene Symbol
col-87
Entrez Gene ID
3565880
Gene Name
COLlagen
Gene Aliases
CELE_Y39G8C.4
Chromosome Location
Chr.II: 14089851 - 14094690 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027262.3 NP_001022433.2 4 - 5 668 73

View Full Product Details