Assay Design



Gene Transcripts

Gene Symbol
sre-46
Entrez Gene ID
3565496
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_Y54G9A.10
Chromosome Location
Chr.II: 13718165 - 13721318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027306.2 NP_001022477.1 2 - 3 653 68

View Full Product Details