Assay Design



Gene Transcripts

Gene Symbol
lgc-29
Entrez Gene ID
3896858
Gene Name
protein-coding
Gene Aliases
CELE_Y45G5AL.2
Chromosome Location
Chr.V: 4138988 - 4143146 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038424.4 NP_001033513.2 9 - 10 1146 122
NM_001330830.1 NP_001317792.1 9 - 10 1100 122

View Full Product Details