Assay Design
Gene Transcripts
- Gene Symbol
- ilys-5
- Entrez Gene ID
-
180928
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F22A3.6
- Chromosome Location
-
Chr.X: 6533493 - 6536840 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_132543.1 |
|
2 - 2 |
521 |
143 |
View Full Product Details