Assay Design



Gene Transcripts

Gene Symbol
fbxa-76
Entrez Gene ID
4363050
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y119D3B.22
Chromosome Location
Chr.III: 1247618 - 1249184 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381727.1 NP_001368053.1 2 - 3 653 72

View Full Product Details