Assay Design
Gene Transcripts
- Gene Symbol
- clx-1
- Entrez Gene ID
-
177556
- Gene Name
- CoLlagen sequence X-hybridizing
- Gene Aliases
-
CELE_C50F7.2
- Chromosome Location
-
Chr.IV: 7734648 - 7736324 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068868.6 |
NP_501269.2 |
1 - 1 |
1547 |
62 |
View Full Product Details