Assay Design



Gene Transcripts

Gene Symbol
EXTL3
Entrez Gene ID
486091
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.25: 28788803 - 28833964 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410096.1 XP_022265804.1 4 - 5 3005 110

View Full Product Details