Assay Design



Gene Transcripts

Gene Symbol
SMAD7
Entrez Gene ID
490570
Gene Name
SMAD family member 7
Gene Aliases
-
Chromosome Location
Chr.7: 79706485 - 79734673 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623131.2 XP_005623188.1 1 - 2 843 122
XM_845400.4 XP_850493.2 1 - 2 843 122

View Full Product Details