Assay Design



Gene Transcripts

Gene Symbol
EBAG9
Entrez Gene ID
403500
Gene Name
estrogen receptor binding site associated antigen 9
Gene Aliases
RCAS1
Chromosome Location
Chr.13: 10081700 - 10099737 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002996.1 NP_001002996.1 1 - 1 102 142
XM_005627797.3 XP_005627854.1 1 - 1 236 142
XM_005627798.3 XP_005627855.1 1 - 1 311 142
XM_005627799.3 XP_005627856.1 1 - 1 236 142
GenBank mRNA AB083366.1 - 1 - 1 102 142

View Full Product Details