Assay Design



Gene Transcripts

Gene Symbol
CXCL10
Entrez Gene ID
478432
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
-
Chromosome Location
Chr.32: 598406 - 599925 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010949.1 NP_001010949.1 2 - 3 186 69
GenBank mRNA AB183191.1 - 2 - 3 186 69

View Full Product Details