Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003106.1 | NP_001003106.1 | 3 - 4 | 625 | 102 |
GenBank mRNA | AF143204.1 | - | 3 - 4 | 592 | 102 |
AF154843.1 | - | 3 - 4 | 625 | 102 | |
AF215893.1 | - | 3 - 4 | 608 | 102 | |
LC191194.1 | - | 3 - 4 | 592 | 102 |