Assay Design



Gene Transcripts

Gene Symbol
RLN2
Entrez Gene ID
403742
Gene Name
relaxin 2
Gene Aliases
RLN, RLN1, relaxin
Chromosome Location
Chr.1: 93500682 - 93505098 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003132.1 NP_001003132.1 1 - 2 206 98
GenBank mRNA AF233687.1 - 1 - 2 206 98
KC977307.1 - 1 - 2 117 98

View Full Product Details