Assay Design



Gene Transcripts

Gene Symbol
IL13
Entrez Gene ID
442990
Gene Name
interleukin 13
Gene Aliases
-
Chromosome Location
Chr.11: 20957870 - 20961391 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003384.1 NP_001003384.1 3 - 4 340 95
XM_022424907.1 XP_022280615.1 2 - 3 880 95
GenBank mRNA AF244915.1 - 3 - 4 340 95

View Full Product Details