Assay Design



Gene Transcripts

Gene Symbol
FMO3
Entrez Gene ID
403603
Gene Name
flavin containing dimethylaniline monoxygenase 3
Gene Aliases
-
Chromosome Location
Chr.7: 27720420 - 27745522 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003060.2 NP_001003060.1 5 - 6 735 68
GenBank mRNA AF384054.1 - 4 - 5 691 68

View Full Product Details