Assay Design



Gene Transcripts

Gene Symbol
PPP1CC
Entrez Gene ID
403557
Gene Name
protein phosphatase 1 catalytic subunit gamma
Gene Aliases
-
Chromosome Location
Chr.26: 8497350 - 8518467 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003033.1 NP_001003033.1 - 877 80
XM_005636001.3 XP_005636058.1 - 1155 80
GenBank mRNA AF525130.1 - - 877 80

View Full Product Details