Assay Design



Gene Transcripts

Gene Symbol
CE10
Entrez Gene ID
403792
Gene Name
CE10 protein
Gene Aliases
-
Chromosome Location
Chr.24: 32847081 - 32848913 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003165.1 NP_001003165.1 1 - 2 130 89
GenBank mRNA AJ238953.1 - 1 - 2 130 89

View Full Product Details