Assay Design



Gene Transcripts

Gene Symbol
TMEM47
Entrez Gene ID
403572
Gene Name
transmembrane protein 47
Gene Aliases
BCMP1
Chromosome Location
Chr.X: 29675386 - 29705941 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003045.1 NP_001003045.1 1 - 2 422 78
GenBank mRNA AJ243320.1 - 1 - 2 422 78

View Full Product Details