Assay Design



Gene Transcripts

Gene Symbol
ID3
Entrez Gene ID
403547
Gene Name
inhibitor of DNA binding 3, HLH protein
Gene Aliases
-
Chromosome Location
Chr.2: 75880659 - 75881786 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003025.2 NP_001003025.1 2 - 3 447 75
GenBank mRNA AJ271644.1 - 2 - 3 459 75

View Full Product Details