Assay Design



Gene Transcripts

Gene Symbol
LHB
Entrez Gene ID
403959
Gene Name
luteinizing hormone subunit beta
Gene Aliases
LH, LH-B, LSH-B
Chromosome Location
Chr.1: 107382855 - 107384029 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197033.1 NP_001183962.1 1 - 2 181 90
GenBank mRNA Y00518.1 - - 174 90

View Full Product Details