Assay Design



Gene Transcripts

Gene Symbol
B3GAT2
Entrez Gene ID
481875
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.12: 33321691 - 33400516 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013419.1 NP_001013437.1 3 - 4 903 85
GenBank mRNA AJ888982.1 - 3 - 4 903 85

View Full Product Details