Assay Design



Gene Transcripts

Gene Symbol
KITLG
Entrez Gene ID
403507
Gene Name
KIT ligand
Gene Aliases
710-712, CSF, MGF
Chromosome Location
Chr.15: 29591068 - 29671985 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012735.2 NP_001012753.1 3 - 4 385 111
XM_022427652.1 XP_022283360.1 3 - 4 396 111
GenBank mRNA AY094361.1 - 2 - 3 144 111
S53329.1 - 2 - 3 191 111

View Full Product Details