Assay Design



Gene Transcripts

Gene Symbol
KITLG
Entrez Gene ID
403507
Gene Name
KIT ligand
Gene Aliases
710-712, CSF, MGF
Chromosome Location
Chr.15: 29591068 - 29671985 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012735.2 NP_001012753.1 5 - 6 722 115
GenBank mRNA AY094361.1 - 4 - 5 481 115
S53329.1 - 4 - 5 528 115

View Full Product Details