Assay Design



Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
403534
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
-
Chromosome Location
Chr.30: 32741582 - 32778883 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003013.2 NP_001003013.2 1 - 2 55 74
GenBank mRNA AY162293.1 - 1 - 2 55 74

View Full Product Details