Assay Design



Gene Transcripts

Gene Symbol
KLRA1
Entrez Gene ID
403478
Gene Name
killer cell lectin-like receptor subfamily A, member 1
Gene Aliases
LY49
Chromosome Location
Chr.27: 35144714 - 35153689 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002986.1 NP_001002986.1 4 - 5 570 105
GenBank mRNA AY191818.1 - 4 - 5 570 105

View Full Product Details