Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002991.1 | NP_001002991.1 | 1 - 2 | 88 | 113 |
GenBank mRNA | AY264843.1 | - | 1 - 2 | 88 | 113 |
EF591860.1 | - | 1 - 2 | 72 | 113 | |
EF591861.1 | - | 1 - 2 | 72 | 113 | |
EF641293.1 | - | 1 - 2 | 72 | 113 |