Assay Design



Gene Transcripts

Gene Symbol
KRT2
Entrez Gene ID
444850
Gene Name
keratin 2
Gene Aliases
CK-2e, K2E, KRT2A
Chromosome Location
Chr.27: 2447854 - 2454492 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003386.2 NP_001003386.2 8 - 9 1546 70
GenBank mRNA AY318943.1 - 8 - 9 1540 70

View Full Product Details