Assay Design



Gene Transcripts

Gene Symbol
KRT1
Entrez Gene ID
444857
Gene Name
keratin 1
Gene Aliases
KER1
Chromosome Location
Chr.27: 2422150 - 2427160 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003392.1 NP_001003392.1 2 - 3 803 68
GenBank mRNA AY318945.1 - 2 - 3 803 68

View Full Product Details