Assay Design



Gene Transcripts

Gene Symbol
KRT1
Entrez Gene ID
444857
Gene Name
keratin 1
Gene Aliases
KER1
Chromosome Location
Chr.27: 2422150 - 2427160 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003392.1 NP_001003392.1 4 - 5 969 76
GenBank mRNA AY318945.1 - 4 - 5 969 76

View Full Product Details