Assay Design



Gene Transcripts

Gene Symbol
MSTN
Entrez Gene ID
403433
Gene Name
myostatin
Gene Aliases
GDF8
Chromosome Location
Chr.37: 729172 - 734255 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002959.1 NP_001002959.1 2 - 3 746 109
GenBank mRNA AY367768.1 - 2 - 3 746 109

View Full Product Details