Assay Design



Gene Transcripts

Gene Symbol
EMR4
Entrez Gene ID
611665
Gene Name
EGF-like module containing, mucin-like, hormone receptor-like sequence 4
Gene Aliases
EMR4P
Chromosome Location
Chr.20: 53342233 - 53385263 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038665.2 NP_001033754.2 7 - 8 657 126
XM_005632346.3 XP_005632403.1 8 - 9 788 126
GenBank mRNA DQ227278.1 - 7 - 8 657 126

View Full Product Details