Assay Design



Gene Transcripts

Gene Symbol
PNLIPRP1
Entrez Gene ID
404010
Gene Name
pancreatic lipase related protein 1
Gene Aliases
PLRP1
Chromosome Location
Chr.28: 27167754 - 27183104 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003319.2 NP_001003319.2 7 - 8 718 58
GenBank mRNA DN356885.1 - 3 - 4 252 58
M35302.1 - 7 - 8 724 58

View Full Product Details