Assay Design



Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
442981
Gene Name
CD3e molecule
Gene Aliases
-
Chromosome Location
Chr.5: 15403724 - 15415259 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003379.1 NP_001003379.1 - 793 88
GenBank mRNA M55410.1 - - 793 88

View Full Product Details