Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003303.1 | NP_001003303.1 | 2 - 3 | 267 | 106 |
XM_022410845.1 | XP_022266553.1 | 2 - 3 | 840 | 106 | |
XM_022410846.1 | XP_022266554.1 | 3 - 4 | 725 | 106 | |
XM_022410847.1 | XP_022266555.1 | 2 - 3 | 674 | 106 | |
XM_022410848.1 | XP_022266556.1 | 3 - 4 | 559 | 106 | |
XM_022410849.1 | XP_022266557.1 | 2 - 3 | 458 | 106 | |
GenBank mRNA | U15593.1 | - | 2 - 3 | 267 | 106 |