Assay Design



Gene Transcripts

Gene Symbol
FUCA1
Entrez Gene ID
403929
Gene Name
alpha-L-fucosidase 1
Gene Aliases
-
Chromosome Location
Chr.2: 75665473 - 75677087 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003250.1 NP_001003250.1 5 - 6 986 60
GenBank mRNA U29765.1 - 5 - 6 986 60
X92448.1 - - 986 60

View Full Product Details