Assay Design



Gene Transcripts

Gene Symbol
VCAM1
Entrez Gene ID
403982
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.6: 49341669 - 49359841 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003298.1 NP_001003298.1 1 - 2 156 86
XM_005621302.3 XP_005621359.2 1 - 2 225 86
GenBank mRNA U32086.1 - 1 - 2 156 86

View Full Product Details