Assay Design



Gene Transcripts

Gene Symbol
TMED11
Entrez Gene ID
403969
Gene Name
glycoprotein 25L
Gene Aliases
GP25L
Chromosome Location
Chr.3: 91435697 - 91452352 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003286.1 NP_001003286.1 2 - 3 226 103
GenBank mRNA X53592.1 - - 226 103

View Full Product Details