Assay Design



Gene Transcripts

Gene Symbol
ACTG1
Entrez Gene ID
475923
Gene Name
actin gamma 1
Gene Aliases
ACTA1
Chromosome Location
Chr.9: 635978 - 638328 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003349.2 NP_001003349.2 - 934 105
GenBank mRNA DQ131478.1 - 1 - 1 271 105

View Full Product Details