Assay Design
Gene Transcripts
- Gene Symbol
- BST2
- Entrez Gene ID
-
609654
- Gene Name
- bone marrow stromal cell antigen 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 45383757 - 45387420 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_860510.5 |
XP_865603.1 |
2 - 3 |
547 |
143 |
View Full Product Details