Assay Design



Gene Transcripts

Gene Symbol
SHOC2
Entrez Gene ID
477819
Gene Name
SHOC2 leucine rich repeat scaffold protein
Gene Aliases
-
Chromosome Location
Chr.28: 22241886 - 22338418 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637720.3 XP_005637777.1 8 - 9 1977 109
XM_022411763.1 XP_022267471.1 7 - 8 1733 109
XM_535013.6 XP_535013.2 8 - 9 1871 109

View Full Product Details