Assay Design
Gene Transcripts
- Gene Symbol
- SH2D1A
- Entrez Gene ID
-
612899
- Gene Name
- SH2 domain containing 1A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 95739568 - 95762117 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_847029.5 |
XP_852122.1 |
2 - 3 |
412 |
145 |
View Full Product Details