Assay Design



Gene Transcripts

Gene Symbol
KCNIP2
Entrez Gene ID
403598
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
Chromosome Location
Chr.28: 14409526 - 14425641 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197190.1 NP_001184119.1 1 - 2 281 81
GenBank mRNA DN373942.1 - 1 - 2 289 81

View Full Product Details