Assay Design
Gene Transcripts
- Gene Symbol
- AHCY
- Entrez Gene ID
-
477198
- Gene Name
- adenosylhomocysteinase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.24: 23410553 - 23423494 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_534388.6 |
XP_534388.2 |
7 - 8 |
1102 |
113 |
View Full Product Details