Assay Design



Gene Transcripts

Gene Symbol
CALM3
Entrez Gene ID
476428
Gene Name
calmodulin 3
Gene Aliases
-
Chromosome Location
Chr.1: 109284729 - 109294491 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022424705.1 XP_022280413.1 3 - 4 267 67
XM_533635.6 XP_533635.1 3 - 4 396 67

View Full Product Details